Learn More
Biomatik Corporation Human Proto-Oncogene Tyrosine-Protein Kinase Src (Src) Elisa Kit
Supplier:  Biomatik Corporation EKC35289
Encompass_Preferred
Human Proto-Oncogene Tyrosine-Protein Kinase Src (Src) Elisa Kit for detecting V-Src Sarcoma (Schmidt-Ruppin A-2) Viral Oncogene Homolog (Avian) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12931; SRC. Target Synonyms: ASV; Avian sarcoma virus; AW259666; c SRC; CDNA FLJ14219 fis clone NT2RP3003800 highly similar to Rattus norvegicus tyrosine protein kinase pp60 c src mRNA; cSrc; EC 2.7.10.2; Neuronal CSRC tyrosine specific protein kinase; Neuronal proto-oncogene tyrosine-protein kinase Src; Neuronal SRC; Oncogene SRC; OTTHUMP00000174476; OTTHUMP00000174477; p60 Src; p60-Src; p60c-src; p60Src; pp60c src; pp60c-src; pp60csrc; Proto oncogene tyrosine protein kinase Src. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4276