Learn More
Biomatik Corporation Human Renalase/ Monoamine Oxidase-C (Rnls/ Mao-C) Elisa Kit
Supplier:  Biomatik Corporation EKC35365
Encompass_Preferred
Human Renalase/ Monoamine Oxidase-C (Rnls/ Mao-C) Elisa Kit for detecting Chromosome 10 Open Reading Frame 59 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ug/ml. Sensitivity: 0.156ug/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5VYX0; RNLS. Target Synonyms: 6530404N21Rik; AI452315; AW060440; C10orf59; Chromosome 10 open reading frame 59; FLJ11218; HGNC:25641; Hypothetical protein LOC55328; MAO C; MAO-C; mMAO C; Monoamine oxidase C; Monoamine oxidase-C; Renalase; Renalase FAD dependent amine oxidase; RNLS; RNLS_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4283