Learn More
Biomatik Corporation Human Secretoglobin Family 1D Member 2 (Scgb1D2) Elisa Kit
Supplier:  Biomatik Corporation EKC35471
Encompass_Preferred
Human Secretoglobin Family 1D Member 2 (Scgb1D2) Elisa Kit for detecting Secretoglobin, Family 1D, Member 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000ng/ml. Sensitivity: 7.81ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95969; SCGB1D2. Target Synonyms: SCGB1D2; LIPHB; LPNBSecretoglobin family 1D member 2; Lipophilin-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4292