Learn More
Biomatik Corporation Human Semaphorin-3G (Sema3G) Elisa Kit
Supplier:  Biomatik Corporation EKC35493
Encompass_Preferred
Human Semaphorin-3G (Sema3G) Elisa Kit for detecting Sema Domain, Immunoglobulin Domain (Ig), Short Basic Domain, Secreted, (Semaphorin) 3G in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NS98; SEMA3G. Target Synonyms: FLJ00014; MGC119473; Sem2; SEM3G_HUMAN; Sema domain immunoglobulin domain (Ig) short basic domain secreted (semaphorin) 3G; Sema3g; Semaphorin 3G; Semaphorin sem2; Semaphorin-3G. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4295