Learn More
Biomatik Corporation Human Sparc-Related Modular Calcium-Binding Protein 2 (Smoc2) Elisa Kit
Catalog No. 502294307
Encompass_Preferred
Human Sparc-Related Modular Calcium-Binding Protein 2 (Smoc2) Elisa Kit for detecting Sparc Related Modular Calcium Binding 2 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H3U7; SMOC2. Target Synonyms: bA270C4A.1; bA37D8.1 (secreted modular calcium binding protein 2); bA37D8.1; dJ421D16.1 (novel thyroglobulin type 1 repeat containing protein); dJ421D16.1; MST117; MSTP117; MSTP140; Secreted modular calcium binding protein 2; Secreted modular calcium-binding protein 2; SMAP 2; SMAP-2; SMAP2; SMOC 2; SMOC-2; SMOC2; SMOC2_HUMAN; Smooth muscle associated protein 2; Smooth muscle-associated protein 2; SPARC related modular calcium binding 2; SPARC-related modular calcium-binding protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-4307 Supplier Biomatik Corporation Supplier No. EKC35641
Human Sparc-Related Modular Calcium-Binding Protein 2 (Smoc2) Elisa Kit for detecting Sparc Related Modular Calcium Binding 2 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H3U7; SMOC2. Target Synonyms: bA270C4A.1; bA37D8.1 (secreted modular calcium binding protein 2); bA37D8.1; dJ421D16.1 (novel thyroglobulin type 1 repeat containing protein); dJ421D16.1; MST117; MSTP117; MSTP140; Secreted modular calcium binding protein 2; Secreted modular calcium-binding protein 2; SMAP 2; SMAP-2; SMAP2; SMOC 2; SMOC-2; SMOC2; SMOC2_HUMAN; Smooth muscle associated protein 2; Smooth muscle-associated protein 2; SPARC related modular calcium binding 2; SPARC-related modular calcium-binding protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.