Learn More
Biomatik Corporation Human T-Complex Protein 1 Subunit Zeta (Cct6A) Elisa Kit
Supplier:  Biomatik Corporation EKC35732
Encompass_Preferred
Human T-Complex Protein 1 Subunit Zeta (Cct6A) Elisa Kit for detecting Chaperonin Containing Tcp1, Subunit 6A (Zeta 1) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40227; CCT6A. Target Synonyms: CCT6A; CCT6; CCTZ; T-complex protein 1 subunit zeta; TCP-1-zeta; Acute morphine dependence-related protein 2; CCT-zeta-1; HTR3; Tcp20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4314