Learn More
Biomatik Corporation Human Thioredoxin Domain-Containing Protein 5 (Txndc5) Elisa Kit
Supplier:  Biomatik Corporation EKC35757
Encompass_Preferred
Human Thioredoxin Domain-Containing Protein 5 (Txndc5) Elisa Kit for detecting Thioredoxin Domain Containing 5 (Endoplasmic Reticulum) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8NBS9; TXNDC5. Target Synonyms: EndoPDI; Endoplasmic reticulum protein ERp46; Endoplasmic reticulum resident protein 46; Endothelial protein disulphide isomerase; ER protein 46; ERp46; Hcc 2; MGC3178; PDIA15; Protein disulfide isomerase family A member 15; STRF8; Thioredoxin domain containing 5 (endoplasmic reticulum); Thioredoxin domain containing 5; Thioredoxin domain containing protein 5; Thioredoxin domain-containing protein 5; Thioredoxin like protein p46; Thioredoxin related protein; TLP46; TXND5_HUMAN; TXNDC 5; Txndc5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4317