Learn More
Biomatik Corporation Human Threonyl-tRNA synthetase, cytoplasmic(TARS) ELISA kit
Supplier:  Biomatik Corporation EKC35765
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156ng/ml- 10 ng/ml. Sensitivity: 0.039 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P26639. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4319