Learn More
Biomatik Corporation Human vascular endothelial cell growth factor E,VEGF-E ELISA Kit
Supplier:  Biomatik Corporation EKC36014
Encompass_Preferred
Sample Type: serum, urine, cell culture supernates, tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25 pg/mL-400 pg/mL. Sensitivity: 1.56 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9NRA1. Target Synonyms: FALLOTEIN, SCDGF, platelet-derived growth factor C|secretory growth factor-like protein|spinal cord-derived growth factor. Restriction: For Research Use Only.
Catalog No. 50-229-4333