Learn More
Biomatik Corporation Human Vasohibin-1 (Vash1) Elisa Kit
Supplier:  Biomatik Corporation EKC36020
Encompass_Preferred
Human Vasohibin-1 (Vash1) Elisa Kit for detecting Vasohibin 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q7L8A9; VASH1. Target Synonyms: KIAA1036; VASH; VASH1; VASH1_HUMAN; Vasohibin 1; Vasohibin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4336