Learn More
Biomatik Corporation Monkey adiponectin (ADP) ELISA kit
Supplier:  Biomatik Corporation EKC36128
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 0.45 ng/mL-30 ng/mL. Sensitivity: 0.11 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4347