Learn More
Biomatik Corporation Monkey gastrin (GAST) ELISA kit
Supplier:  Biomatik Corporation EKC36159
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P33714. Target Synonyms: GAS. Restriction: For Research Use Only.
Catalog No. 50-229-4352