Learn More
Biomatik Corporation Monkey Macrophage Inflammatory Protein-3α(MIP-3α/CCL20)ELISA Kit
Supplier:  Biomatik Corporation EKC36186
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: CKb4, LARC, MIP-3a, MIP3A, SCYA20, ST38, exodus-1|macrophage inflammatory protein 3 alpha|small inducible cytokine subfamily A (Cys-Cys), member 20. Restriction: For Research Use Only.
Catalog No. 50-229-4354