Learn More
Biomatik Corporation Monkey thyroid stimulating hormone(TSH) ELISA kit
Supplier:  Biomatik Corporation EKC36200
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 0.3 μIU/mL-12 μIU/mL. Sensitivity: 0.15 μIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9BEH3. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4356