Learn More
Biomatik Corporation Mouse ADAMTS-like protein 4(ADAMTSL4) ELISA kit
Supplier:  Biomatik Corporation EKC36258
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q80T21. Target Synonyms: TSRC1, OTTHUMP00000032789|thrombospondin repeat containing 1. Restriction: For Research Use Only.
Catalog No. 50-229-4360