Learn More
Biomatik Corporation Mouse Biglycan(BGN) ELISA kit
Supplier:  Biomatik Corporation EKC36409
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P28653. Target Synonyms: DSPG1, PG-S1, PGI, SLRR1A, biglycan proteoglycan|bone/cartilage proteoglycan-I|dermatan sulphate proteoglycan I|small leucine-rich protein 1A. Restriction: For Research Use Only.
Catalog No. 50-229-4370