Learn More
Biomatik Corporation Mouse Carbonic anhydrase 3(CA3) ELISA kit
Supplier:  Biomatik Corporation EKC36464
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 4.69 pg/mL-300 pg/mL. Sensitivity: 1.17 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P16015. Target Synonyms: CAIII, Car3, carbonic anhydrase III. Restriction: For Research Use Only.
Catalog No. 50-229-4375