Learn More
Biomatik Corporation Mouse Carnitine O-Palmitoyltransferase 1, Liver Isoform (Cpt1A) Elisa Kit
Supplier:  Biomatik Corporation EKC36477
Encompass_Preferred
Mouse Carnitine O-Palmitoyltransferase 1, Liver Isoform (Cpt1A) Elisa Kit for detecting Carnitine Palmitoyltransferase 1A (Liver) in serum, plasma, cell lysates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P97742; Cpt1a. Target Synonyms: Cpt1a; Cpt-1; Cpt1; Carnitine O-palmitoyltransferase 1; liver isoform; CPT1-L; Carnitine O-palmitoyltransferase I; liver isoform; CPT I; CPTI-L; Carnitine palmitoyltransferase 1A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4377