Learn More
Biomatik Corporation Mouse Cubilin (Cubn) Elisa Kit
Supplier:  Biomatik Corporation EKC36686
Encompass_Preferred
Mouse Cubilin (Cubn) Elisa Kit for detecting Cubilin (Intrinsic Factor-Cobalamin Receptor) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9JLB4; Cubn. Target Synonyms: Cubn; Ifcr; Cubilin; Intrinsic factor-cobalamin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4393