Learn More
Biomatik Corporation Mouse cytotoxic T lymphocyte associated antigen 4,CTLA-4 ELISA Kit
Supplier:  Biomatik Corporation EKC36715
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09793. Target Synonyms: CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, ICOS, IDDM12, celiac disease 3|cytotoxic T-lymphocyte antigen 4|cytotoxic T-lymphocyte-associated antigen 4|cytotoxic T-lymphocyte-associated serine esterase-4. Restriction: For Research Use Only.
Catalog No. 50-229-4397