Learn More
Biomatik Corporation Mouse Ectonucleotide pyrophosphatase/phosphodiesterase family member 1(ENPP1) ELISA kit
Supplier:  Biomatik Corporation EKC36778
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P06802. Target Synonyms: M6S1, NPP1, NPPS, PC-1, PCA1, PDNP1, Ly-41 antigen|alkaline phosphodiesterase 1|membrane component, chromosome 6, surface marker 1|phosphodiesterase I/nucleotide pyrophosphatase 1|plasma-cell membra. Restriction: For Research Use Only.
Catalog No. 50-229-4401