Learn More
Biomatik Corporation Mouse Fibroblast growth factor 5(FGF5) ELISA kit
Supplier:  Biomatik Corporation EKC36871
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P15656. Target Synonyms: HBGF-5, Smag-82, heparin-binding growth factor 5. Restriction: For Research Use Only.
Catalog No. 50-229-4408