Learn More
Biomatik Corporation Mouse Galanin-like peptide(GALP) ELISA kit
Supplier:  Biomatik Corporation EKC36915
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 23.5 pg/mL-1500 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q810H5. Target Synonyms: alarin|gal-like peptide. Restriction: For Research Use Only.
Catalog No. 50-229-4411