Learn More
Biomatik Corporation Mouse Interferon Alpha-2 (Ifna2) Elisa Kit
Supplier:  Biomatik Corporation EKC37134
Encompass_Preferred
Mouse Interferon Alpha-2 (Ifna2) Elisa Kit for detecting Interferon Alpha-2(Ifna2) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 9.38-600pg/ml. Sensitivity: 2.35pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01573; Ifna2. Target Synonyms: Ifna2Interferon alpha-2; IFN-alpha-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4422