Learn More
Biomatik Corporation Mouse Leukemia inhibitory factor,LIF ELISA kit
Supplier:  Biomatik Corporation EKC37279
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09056. Target Synonyms: CDF, DIA, HILDA, D factor|cholinergic differentiation factor|differentiation inhibitory activity|differentiation stimulating factor. Restriction: For Research Use Only.
Catalog No. 50-229-4426