Learn More
Biomatik Corporation Mouse lymphotactin,Lptn/LTN ELISA Kit
Supplier:  Biomatik Corporation EKC37318
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 ng/ml - 10 ng/ml. Sensitivity: 0.039 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P47993. Target Synonyms: ATAC, LPTN, LTN, SCM-1, SCM-1a, SCM1, SCYC1, lymphotactin|small inducible cytokine subfamily C, member 1 (lymphotactin). Restriction: For Research Use Only.
Catalog No. 50-229-4429