Learn More
Biomatik Corporation Mouse Melanoma-derived growth regulatory protein(MIA) ELISA kit
Supplier:  Biomatik Corporation EKC37377
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 4.7 pg/mL-300 pg/mL. Sensitivity: 1.17 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q61865. Target Synonyms: CD-RAP. Restriction: For Research Use Only.
Catalog No. 50-229-4439