Learn More
Biomatik Corporation Mouse Peroxisome Proliferator-activated receptor γ,PPAR-γ ELISA Kit
Supplier:  Biomatik Corporation EKC37563
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.625 pg/mL-40 pg/mL. Sensitivity: 0.156 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P37238. Target Synonyms: CIMT1, GLM1, NR1C3, PPARG1, PPARG2, PPARgamma, OTTHUMP00000185030|OTTHUMP00000185033|PPAR gamma|nuclear receptor subfamily 1 group C member 3|peroxisome proliferative activated receptor gamma|peroxi. Restriction: For Research Use Only.
Catalog No. 50-229-4451