Learn More
Biomatik Corporation Mouse Plastin-2 (Lcp1) Elisa Kit
Supplier:  Biomatik Corporation EKC37590
Encompass_Preferred
Mouse Plastin-2 (Lcp1) Elisa Kit for detecting Lymphocyte Cytosolic Protein 1 (L-Plastin) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q61233; Lcp1. Target Synonyms: Lcp1; Pls2; Plastin-2; 65 kDa macrophage protein; L-plastin; Lymphocyte cytosolic protein 1; LCP-1; pp65. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4454