Learn More
Biomatik Corporation Mouse Protein delta homolog 1(DLK1) ELISA kit
Supplier:  Biomatik Corporation EKC37653
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q09163. Target Synonyms: DLK, FA1, PREF1, Pref-1, ZOG, pG2, delta-like 1 homolog|delta-like homolog|fetal antigen 1|preadipocyte factor 1|secredeltin. Restriction: For Research Use Only.
Catalog No. 50-229-4460