Learn More
Biomatik Corporation Mouse Transforming Growth Factor Beta Receptor Type 3 (Tgfbr3) Elisa Kit
Supplier:  Biomatik Corporation EKC37947
Encompass_Preferred
Mouse Transforming Growth Factor Beta Receptor Type 3 (Tgfbr3) Elisa Kit for detecting Transforming Growth Factor, Beta Receptor Iii in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O88393; Tgfbr3. Target Synonyms: Tgfbr3; Transforming growth factor beta receptor type 3; TGF-beta receptor type 3; TGFR-3; Betaglycan; Transforming growth factor beta receptor III; TGF-beta receptor type III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4478