Learn More
Biomatik Corporation Mouse Tumor necrosis factor receptor superfamily member 17(TNFRSF17) ELISA kit
Supplier:  Biomatik Corporation EKC37989
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.39 ng/ml- 20ng/ml. Sensitivity: 0.009 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O88472. Target Synonyms: BCM, BCMA, CD269, B cell maturation antigen|B-cell maturation factor. Restriction: For Research Use Only.
Catalog No. 50-229-4481