Learn More
Biomatik Corporation Mouse Vascular endothelial cell growth factor receptor 1,VEGFR-1/Flt1 ELISA kit
Supplier:  Biomatik Corporation EKC38029
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P35969. Target Synonyms: FLT, VEGFR1, fms-related tyrosine kinase 1|vascular endothelial growth factor receptor 1. Restriction: For Research Use Only.
Catalog No. 50-229-4485