Learn More
Biomatik Corporation Mouse WAP four-disulfide core domain protein 2(WFDC2) ELISA kit
Supplier:  Biomatik Corporation EKC38056
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9DAU7. Target Synonyms: EDDM4, HE4, MGC57529, WAP5, dJ461P17.6, WAP domain containing protein HE4-V4|epididymal protein 4|epididymal secretory protein E4|epididymis-specific, whey-acidic protein type, four-disulfide core|m. Restriction: For Research Use Only.
Catalog No. 50-229-4487