Learn More
Biomatik Corporation New Delhi metallo-beta-lactamase-1 (NDM-1) ELISA kit
Supplier:  Biomatik Corporation EKC38078
Encompass_Preferred
Sample Type: serum, plasma, urine samples infected by New Delhi super bacteria. Species Reactivity: Not Available. Detection Range: 6.25 ng/mL- 400 ng/mL. Sensitivity: 3.12 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4490