Learn More
Biomatik Corporation Pig Complement 3,C3 ELISA Kit
Supplier:  Biomatik Corporation EKC38145
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 0.625 μg/mL-10 μg/mL. Sensitivity: 0.625 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01025. Target Synonyms: AHUS5, ARMD9, ASP, CPAMD1, acylation-stimulating protein cleavage product|complement component C3. Restriction: For Research Use Only.
Catalog No. 50-229-4498