Learn More
Biomatik Corporation Pig Keratinocyte Growth Factor,KGF ELISA Kit
Supplier:  Biomatik Corporation EKC38260
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Sus scrofa (Pig). Detection Range: 3.12 pg/mL-200pg/mL. Sensitivity: 0.78 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9N198. Target Synonyms: HBGF-7, KGF, fibroblast growth factor 7|heparin-binding growth factor 7|keratinocyte growth factor. Restriction: For Research Use Only.
Catalog No. 50-229-4507