Learn More
Biomatik Corporation Pig Scavenger receptor cysteine-rich type 1 protein M130(CD163) ELISA kit
Supplier:  Biomatik Corporation EKC38328
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 93.75 ng/mL-6000 ng/mL. Sensitivity: 23.4 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q2VL90. Target Synonyms: M130, MM130, CD163 antigen|macrophage-associated antigen. Restriction: For Research Use Only.
Catalog No. 50-229-4514