Learn More
Biomatik Corporation Rabbit Collagen Type Ⅱ,Col Ⅱ ELISA Kit
Supplier:  Biomatik Corporation EKC38416
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 6.25 ng/mL-400 ng/mL. Sensitivity: 1.56 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Collagen Type 2,Col 2,_x000D_,Collagen Type II,Col II. Restriction: For Research Use Only.
Catalog No. 50-229-4516