Learn More
Biomatik Corporation Rabbit Epidermal growth factor,EGF ELISA Kit
Supplier:  Biomatik Corporation EKC38435
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 0.5 ng/mL-8 ng/mL. Sensitivity: 0.25 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: G1TBJ6. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4520