Learn More
Biomatik Corporation Rabbit follicle-stimulating hormone,FSH ELISA Kit
Supplier:  Biomatik Corporation EKC38443
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 1.56 mIU/mL-100 mIU/mL. Sensitivity: 0.39 mIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6IY73. Target Synonyms: follicle-stimulating hormone beta subunit|follitropin, beta chain, FSHB. Restriction: For Research Use Only.
Catalog No. 50-229-4522