Learn More
Biomatik Corporation Rabbit intercellular adhesion molecule 1,ICAM-1/CD54 ELISA Kit
Supplier:  Biomatik Corporation EKC38460
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: U3KPG6. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4525