Learn More
Biomatik Corporation Rabbit Myeloperoxidase (Mpo) Elisa Kit
Supplier:  Biomatik Corporation EKC38499
Encompass_Preferred
Rabbit Myeloperoxidase (Mpo) Elisa Kit for detecting Myeloperoxidase, Mpo in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 1.25-20ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: G1SR65. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4530