Learn More
Biomatik Corporation Rabbit Pulmonary surfactant-associated protein A(SFTPA1) ELISA kit
Supplier:  Biomatik Corporation EKC38532
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P12842. Target Synonyms: AC068139.6, MGC133365, PSAP, PSPA, SFTP1, SFTPA1, pulmonary surfactant apoprotein|surfactant, pulmonary-associated protein A1|surfactant, pulmonary-associated protein A1B. Restriction: For Research Use Only.
Catalog No. 50-229-4538