Learn More
Biomatik Corporation Rat Apolipoprotein M(APOM) ELISA kit
Supplier:  Biomatik Corporation EKC38704
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P14630. Target Synonyms: DADB-127H9.5, G3a, HSPC336, MGC22400, NG20, NG20-like protein|alternative name: G3a, NG20. Restriction: For Research Use Only.
Catalog No. 50-229-4552