Learn More
Biomatik Corporation Rat glycosaminoglycan/mucopolysaccharide ELISA kit
Supplier:  Biomatik Corporation EKC39133
Encompass_Preferred
Sample Type: serum, plasma, urine. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 6.25 ng/mL-100 ng/mL. Sensitivity: 3.125 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4582