Learn More
Biomatik Corporation Rat growth differentiation factor 15(GDF15)ELISA Kit
Supplier:  Biomatik Corporation EKC39146
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9Z0J6. Target Synonyms: GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB, NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1|PTGF-beta|prostate differentiation factor. Restriction: For Research Use Only.
Catalog No. 50-229-4584