Learn More
Biomatik Corporation Rat low density lipoprotein receptor,LDLR ELISA Kit
Supplier:  Biomatik Corporation EKC39352
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P35952. Target Synonyms: FH, FHC, LDLCQ2, LDL receptor|low-density lipoprotein receptor class A domain-containing protein 3. Restriction: For Research Use Only.
Catalog No. 50-229-4603