Learn More
Biomatik Corporation Rat Monoamine Oxidase (Mao) Elisa Kit
Supplier:  Biomatik Corporation EKC39412
Encompass_Preferred
Rat Monoamine Oxidase (Mao) Elisa Kit for detecting Monoamine Oxidase(Mao) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 93.75-6000mU/ml. Sensitivity: 23.43mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21396; Maoa. Target Synonyms: Maoa; Amine oxidase [flavin-containing] A; Monoamine oxidase type A; MAO-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4609