Learn More
Biomatik Corporation Rat plasmin-antiplasmin complex,PAP ELISA Kit
Supplier:  Biomatik Corporation EKC39553
Encompass_Preferred
Sample Type: serum, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 7.8 ng/mL-500 ng/mL. Sensitivity: 1.95 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4625